missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear New York S1P Shoe

Product Code. 17562466
Click to view available options
Size:
35
36
37
38
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17562466

Brand: Emma Safety Footwear MM43264748

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Breathable knitted textile for optimal comfort and great performance. With the New York safety shoe, you get optimal protection while looking good.

  • A highly sporty shoe with optimum performance in the field of safety. New York is the shoe that combines good looks with maximum protection.
  • Thanks to the knitted textile upper and the Nano Carbon Composite protective nose, the New York is particularly light and comfortable. Ideal for workplaces where safety and long-lasting comfort are top priorities.
  • The TPU GripForce™ Easy Twist outsole offers maximum grip, is durable and highly recyclable.
  • The integrated lacing in this great shoe prevents pressure points from hooks and eyes and the Hydro-Tec™ Antibacterial lining provides optimum breathability.
  • New York exudes pure sportsmanship and is designed for performance and looks.
  • Industries:
    • Logistics & transport
    • Electronics
    • Manufacturing
  • Safety shoe
    • S1P-Safetyshoe with sole plate for dry work

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black, Gray, Orange
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
Material Polyurethane
Size 48
Toe Material Composite
Fit Standard Width
Ankle Style High
Product Type Safety Shoe
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.