missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Pig High-Visibility Yellow Mats and Rolls
Click to view available options
Capacity (English):
28.16 oz.
Unit Size:
Pack of 100
Description
- Heavyweight mat pads are ideal for soaking up big spills and heavy leaks around machines and equipment; dispenser box fits neatly on shelves or carts
- Bright yellow color stands out so it's easy for workers to see hazards, repairs or cleanups in progress, even in poorly lit areas.
- PIG Mat lasts 2 times longer than competitive mats and won't rip, tear or fray even when saturated; liquids absorb quickly for faster, easier cleanups and safer workplaces
- Absorbs most common industrial liquids - oils, coolants, solvents, water and more
Specifications
Specifications
| Color | Yellow |
| Capacity (English) | 28.16 oz. |
| Capacity (Metric) | 0.83 L |
| Material | Polypropylene |
| Type | High Visibility Absorbent Mat Pad |
| For Use With (Application) | Absorbs oils, coolants, solvents and water |
| Dimensions (L x W) | 20 x 15 in. (50.8 x 38 cm) |
| Length (English) | 20 in. |
| Length (Metric) | 50.8 cm |
| Width (English) | 15 in. |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction