missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Jersey TRPH Standard 987 200 μg/mL in Dichloromethane:Carbon Disulfide (9:1), Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Heneicosane∣n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hentriacontane∣n-Heptacosane∣n-Heptadecane∣n-Heptatriacontane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Nonacosane∣n-Nonadecane∣n-Nonane∣n-Nonatriacontane∣n-Octadecane∣n-Octane∣n-Octatriacontane∣n-Pentacosane∣n-Pentadecane∣n-Pentatriacontane∣n-Tetradecane∣n-Tetratriacontane∣n-Tricosane∣n-Tridecane∣n-Tritriacontane∣n-Undecane∣Octacosane∣Phytane∣Pristane∣Tetracontane∣Tetracosane∣Triacontane |
| CAS | 629-94-7, 124-18-5, 629-97-0, 112-40-3, 544-85-4, 112-95-8 |
| Analyte or Component Names | Heneicosane∣n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hentriacontane∣n-Heptacosane∣n-Heptadecane∣n-Heptatriacontane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Nonacosane∣n-Nonadecane∣n-Nonane∣n-Nonatriacontane∣n-Octadecane∣n-Octane∣n-Octatriacontane∣n-Pentacosane∣n-Pentadecane∣n-Pentatriacontane∣n-Tetradecane∣n-Tetratriacontane∣n-Tricosane∣n-Tridecane∣n-Tritriacontane∣n-Undecane∣Octacosane∣Phytane∣Pristane∣Tetracontane∣Tetracosane∣Triacontane |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane/Carbon Disulfide |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Heneicosane 200 μg/mL∣n-Decane 200 μg/mL∣n-Docosane 200 μg/mL∣n-Dodecane 200 μg/mL∣n-Dotriacontane 200 μg/mL∣n-Eicosane 200 μg/mL∣n-Hentriacontane 200 μg/mL∣n-Heptacosane 200 μg/mL∣n-Heptadecane 200 μg/mL∣n-Heptatriacontane 200 μg/mL∣n-Hexacosane 200 μg/mL∣n-Hexadecane 200 μg/mL∣n-Hexatriacontane 200 μg/mL∣n-Nonacosane 200 μg/mL∣n-Nonadecane 200 μg/mL∣n-Nonane 200 μg/mL∣n-Nonatriacontane 200 μg/mL∣n-Octadecane 200 μg/mL∣n-Octane 200 μg/mL∣n-Octatriacontane 200 μg/mL∣n-Pentacosane 200 μg/mL∣n-Pentadecane 200 μg/mL∣n-Pentatriacontane 200 μg/mL∣n-Tetradecane 200 μg/mL∣n-Tetratriacontane 200 μg/mL∣n-Tricosane 200 μg/mL∣n-Tridecane 200 μg/mL∣n-Tritriacontane 200 μg/mL∣n-Undecane 200 μg/mL∣Octacosane 200 μg/mL∣Phytane 200 μg/mL∣Pristane 200 μg/mL∣Tetracontane 200 μg/mL∣Tetracosane 200 μg/mL∣Triacontane 200 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?