missing translation for 'onlineSavingsMsg'
Learn More

New Fuchsin, MP Biomedicals™

Product Code. 11445021 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445021 100 g 100g
11435021 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445021 Supplier MP Biomedicals Supplier No. 0215582380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Soluble in water and ethanol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3248-91-7
Molecular Formula C22H24ClN3
Molecular Weight (g/mol) 365.91
MDL Number MFCD00012567
InChI Key ORDGVBRMUJCHEO-UHFFFAOYSA-M
Synonym new fuchsin, magenta iii, basic violet 2, neofuchsine, new fuchsine, new magenta, fuchsine sbp, c.i. basic violet 2, c.i. basic violet 2, monohydrochloride, new-fuchsin
PubChem CID 14454445
ChEBI CHEBI:87671
IUPAC Name tris(4-amino-3-methylphenyl)methylium chloride
SMILES [Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1

Specifications

Color Green
Quantity 100 g
Formula Weight 365.91
Physical Form Crystalline Powder
Chemical Name or Material New Fuchsin
compliance-icons
Product Identifier
  • New Fuchsin
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Indicazioni di pericolo
  • H351-Suspected of causing cancer.
Consigli di prudenza
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informazioni supplementari
  • MIXTURE LIST-Contain: New Fuchsin
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.