missing translation for 'onlineSavingsMsg'
Learn More

New Fuchsin, MP Biomedicals™

Product Code. 11435021 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11435021 25 g 25g
11445021 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11435021

Brand: MP Biomedicals 0215582325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Soluble in water and ethanol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3248-91-7
Molecular Formula C22H24ClN3
Molecular Weight (g/mol) 365.91
MDL Number MFCD00012567
InChI Key ORDGVBRMUJCHEO-UHFFFAOYSA-M
Synonym new fuchsin, magenta iii, basic violet 2, neofuchsine, new fuchsine, new magenta, fuchsine sbp, c.i. basic violet 2, c.i. basic violet 2, monohydrochloride, new-fuchsin
PubChem CID 14454445
ChEBI CHEBI:87671
IUPAC Name tris(4-amino-3-methylphenyl)methylium chloride
SMILES [Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1

Specifications

Color Green
Quantity 25 g
Formula Weight 365.91
Physical Form Crystalline Powder
Chemical Name or Material New Fuchsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.