missing translation for 'onlineSavingsMsg'
Learn More

Unity Lab Services™ New Equipment Startup Service Incubators, Ovens, Vacuum Oven And Furnaces 3rd Party

Product Code. 30286561
Change view
Click to view available options
Duration:
1 Day
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration Unit Size
30286561 1 Day Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30286561 Supplier Unity Lab Services™ Supplier No. STARTCOVMVS
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Startup service for new ovens, vacuum ovens, furnaces and incubators not manufactured by Thermo Fisher Scientific. Excludes major assembly.

Startup service for ovens, vacuum ovens, furnaces and incubators manufactured by third parties.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Ovens, Vacuums, Furnaces, Incubators
Duration 1 Day
Product Type Ovens, Vacuums, Furnaces, Incubators
Target Audience User seeking to learn about specific instruments/software features
Species All
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.