Learn More
Eppendorf™ Test Tube Racks for Model C76 Water Bath Shaker
Product Code. 11648613
Click to view available options
Holds:
18.0 25 mm Tubes
29.0 16 mm Tubes
36.0 20 mm Tubes
Unit Size:
Each
Product Code. 11648613
Brand: Eppendorf™ AG7TT16
Only null left
Add to basket
Add to basket
Test Tube Racks have an adjustable angle and tube height support. Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Require subplatform (14-284-46); order separately.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) C76 water bath shaker
Holds 29.0 16 mm Tubes
Includes Insert
Material Stainless Steel