Learn More
Eppendorf™ Scientific Test Tube Racks
Product Code. 11399924
Change view
Click to view available options
Size:
Small
Medium
Diameter (Metric):
12 to 15 mm
15 to 18 mm
18 to 21 mm
22 to 26 mm
26 to 30 mm
8 to 11 mm
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Diameter (Metric) unitSize
11399924 Small 12 to 15 mm Each
11389924 Small 8 to 11 mm Each
11309934 Small 15 to 18 mm Each
11319934 Small 18 to 21 mm Each
11329934 Small 22 to 26 mm Each
11329924 Medium 8 to 11 mm Each
11339924 Medium 12 to 15 mm Each
11349924 Medium 15 to 18 mm Each
11339934 Small 26 to 30 mm Each
11369924 Medium 22 to 26 mm Each
11359924 Medium 18 to 21 mm Each
11379924 Medium 26 to 30 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11399924 Supplier Eppendorf™ Supplier No. M12890002
Only null left
Add to basket
Add to basket
Adjustable angle and tube height
  • Inserts to help retain tubes and prevent them from spinning in the rack
  • Finger-tight knobs to quickly adjust angle, or release the rack from the base plate, for easy transport to the bench
  • Stainless-steel
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Tube Rack
Material Stainless Steel
Diameter (Metric) 12 to 15 mm
For Use With (Equipment) Innova 43/43R Incubator shaker
Includes Two adjustable bars and non-skid rubber mat
No. of Positions 34
Size Small