missing translation for 'onlineSavingsMsg'
Learn More

Nevirapine N10-Oxide, Mikromol™

Product Code. 30051005
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30051005

Brand: Mikromol MM1146.060025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nevirapine N10-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nevirapine N10-Oxide
CAS 1027324-99-7
Analyte or Component Names Nevirapine N10-Oxide
Shipping Condition Room Temperature
Molecular Formula C15 H14 N4 O2
Certifications/Compliance ISO 9001
Synonym 6H-Dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, 11-cyclopropyl-5,11-dihydro-4-methyl-, 10-oxide
InChI Formula InChI=1S/C15H14N4O2/c1-9-6-7-16-13-12(9)17-14(20)11-3-2-8-18(21)15(11)19(13)10-4-5-10/h2-3,6-8,10H,4-5H2,1H3,(H,17,20)
SMILES Cc1ccnc2N(C3CC3)c4c(ccc[n+]4[O-])C(=O)Nc12
IUPAC Name 11-cyclopropyl-4-methyl-10-oxido-5H-dipyrido[3,2-a:2',3'-d][1,4]diazepin-10-ium-6-one
Molecular Weight (g/mol) 282.3
Quantity 25 mg
Formula Weight 282.1117 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.