missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nevada Pesticide Mixture Version 2 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Acequinocyl∣Bifenazate∣Bifenthrin∣Cyfluthrin∣Cypermethrin∣Daminozide∣Dimethomorph∣Etoxazole∣Fenhexamid∣Flonicamid∣Fludioxonil∣Imidacloprid∣Myclobutanil∣Paclobutrazol∣Piperonyl butoxide∣Pyrethrins∣Quintozene∣Spinetoram J∣Spinosad∣Spirotetramat∣Thiamethoxam∣Trifloxystrobin |
| CAS | 71751-41-2, 57960-19-7, 149877-41-8, 82657-04-3, 68359-37-5, 52315-07-8 |
| Analyte or Component Names | Abamectin∣Acequinocyl∣Bifenazate∣Bifenthrin∣Cyfluthrin∣Cypermethrin∣Daminozide∣Dimethomorph∣Etoxazole∣Fenhexamid∣Flonicamid∣Fludioxonil∣Imidacloprid∣Myclobutanil∣Paclobutrazol∣Piperonyl butoxide∣Pyrethrins∣Quintozene∣Spinetoram J∣Spinosad∣Spirotetramat∣Thiamethoxam∣Trifloxystrobin |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Abamectin 100 μg/mL∣Acequinocyl 100 μg/mL∣Bifenazate 100 μg/mL∣Bifenthrin 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Daminozide 100 μg/mL∣Dimethomorph 100 μg/mL∣Etoxazole 100 μg/mL∣Fenhexamid 100 μg/mL∣Flonicamid 100 μg/mL∣Fludioxonil 100 μg/mL∣Imidacloprid 100 μg/mL∣Myclobutanil 100 μg/mL∣Paclobutrazol 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Pyrethrins 100 μg/mL∣Quintozene 100 μg/mL∣Spinetoram J 100 μg/mL∣Spinosad 100 μg/mL∣Spirotetramat 100 μg/mL∣Thiamethoxam 100 μg/mL∣Trifloxystrobin 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction