missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophils Antibody (Nimp-R14), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-80078CL1
This item is not returnable.
View return policy
Description
Neutrophils Monoclonal antibody specifically detects Neutrophils in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Neutralization,Immunocytochemistry/Immunofluorescence
Specifications
| Neutrophils | |
| Monoclonal | |
| CoraFluor 1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization | |
| Nimp-R14 | |
| PBS | |
| Purified mouse BALB/c neutrophils. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction