missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophils Antibody (Nimp-R14), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80078AF700
This item is not returnable.
View return policy
Description
Neutrophils Monoclonal antibody specifically detects Neutrophils in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Neutrophils | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Nimp-R14 | |
| 50mM Sodium Borate | |
| Purified mouse BALB/c neutrophils. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction