missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neutrophil elastase Monoclonal Antibody (ELA10-103.2), Invitrogen™

Mouse Monoclonal Antibody
£367.00 - £916.00
Specifications
| Antigen | Neutrophil elastase |
|---|---|
| Clone | ELA10-103.2 |
| Concentration | 1.28 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17820246
|
Invitrogen™
MA5-46148 |
100 μg |
£367.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11598892
![]() |
Invitrogen™
MA110609 |
500 μg |
£466.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17880196
|
Invitrogen™
MA5-46009 |
1 mg |
£916.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10609 detects Neutrophil elastase from human samples. MA1-10609 has been successfully used in ELISA applications. The MA1-10609 immunogen is purified human Neutrophil Elastase.
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.Specifications
| Neutrophil elastase | |
| 1.28 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease | |
| ELANE | |
| IgG2b | |
| Protein A | |
| Antibody |
| ELA10-103.2 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08246 | |
| 1991 | |
| Purified human Neutrophil Elastase. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| ELANE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title