missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neutrophil elastase Monoclonal Antibody (ELA10-101.5), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110608
This item is not returnable.
View return policy
Description
MA1-10608 detects Neutrophil elastase from human samples. MA1-10608 has been successfully used in ELISA and in studies of inhibition and enzyme kinetics of neutrophil elastase. The MA1-10608 immunogen is purified human Neutrophil Elastase.
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.
Specifications
| Neutrophil elastase | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P08246 | |
| ELANE | |
| Purified human Neutrophil Elastase. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| ELA10-101.5 | |
| Unconjugated | |
| ELANE | |
| bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1991 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction