missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neutrophil elastase Monoclonal Antibody (39A)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183125
This item is not returnable.
View return policy
Description
Mouse anti Human elastase antibody, clone 39A recognizes human neutrophil elastase, also known as elastase-2, Bone marrow serine protease, PMN elastase or Medullasin.
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.
Specifications
| Neutrophil elastase | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with <0.1% sodium azide | |
| P08246 | |
| ELANE | |
| Elastase from human neutrophils. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay | |
| 39A | |
| Unconjugated | |
| ELANE | |
| bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease | |
| Mouse | |
| Protein G | |
| RUO | |
| 1991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction