missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophil Elastase/ELA2 Antibody (2H12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26603-50ul
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Monoclonal antibody specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| Neutrophil Elastase/ELA2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 2H12 | |
| Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
| A synthesized peptide derived from Human Neutrophil Elastase/ELA2 [UniProt P08246] | |
| 50 μL | |
| Cancer, Neuroscience | |
| 1991 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction