missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NeutrAvidin Protein, DyLight™ 550
Click to view available options
Quantity:
1mL
Unit Size:
1mL
Description
NeutrAvidin Protein, DyLight 550 conjugate, 84606, from Invitrogen., 84606, from Invitrogen. Species Reactivity: ; Applications: Immunofluorescence.
Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14ug/mg of protein, which is near the theoretical maximum activity.
Specifications
Specifications
| Concentration | 1mg/mL |
| Conjugate | DyLight 550 |
| Cross Reactivity | Tag |
| Formulation | Buffered saline solution with 1% BSA and no preservative |
| Quantity | 1mL |
| Regulatory Status | RUO |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Immunofluorescence |
| Purity or Quality Grade | purified |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction