missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NeutrAvidin Protein, DyLight™ 405

Product Code. 10095803
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10095803 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10095803 Supplier Invitrogen™ Supplier No. 22831

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein

NeutrAvidin Protein, DyLight 405 conjugate, 22831, from Invitrogen., 22831, from Invitrogen. Species Reactivity: ; Applications: Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14ug/mg of protein, which is near the theoretical maximum activity. 22831 contains 1 mL of protein in clear liquid at a concentration of 1mg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
Conjugate DyLight 405
Cross Reactivity Tag
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.2
pH 7.2
Quantity 1 mL
Regulatory Status RUO
Content And Storage 4°C
Form Liquid
For Use With (Application) Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Purity or Quality Grade purified
Host Species N/A
Product Type NeutrAvidin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.