missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Neutral Red Stain, Concentrated
Click to view available options
Quantity:
100mL
Unit Size:
100mL
Description
Pro-Lab Diagnostics™ stains are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all stains offer excellent performance characteristics.
- 10X concentrated – 100mL is diluted to 1L of ready-to-use stain
- Neutral Red is used as a counterstain to easily identify gram-negative bacteria
- Apply after decolorization to make decolorized organisms visible
Specifications
Specifications
| Solution Type | Concentrated Reagent |
| Type | Neutral Red Stain |
| Quantity | 100mL |
| Certifications/Compliance | CE marked (98/78/EC) |
| Format | Liquid |
| For Use With (Application) | Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria |
| Color | Red |
| Composition | Ethanol 25 – <50%, Glycerol 10 – <25%, Methanol 2.5 – <5%, and N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride 2.5 – <5% |
| Storage Requirements | Keep at temperature not exceeding 20°C |
| Stain Type | Gram Counterstain |
| Show More |
Safety and Handling
Product Identifier
- Neutral Red Concentrate
- Warning
- Flammable liquid Category 3
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- Specific target organ toxicity Category 2
- H226-Flammable liquid and vapour.
- H332-Harmful if inhaled.
- H341-Suspected of causing genetic defects.
- H371-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501-Dispose of contents/container to .
- P202-Do not handle until all safety precautions have been read and understood.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P370+P378-In case of fire: Use to extinguish.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P405-Store locked up.
- MIXTURE LIST-Contains : ethanol, glycerol, N8, N8, 3-trimethyl-2, 8-phenazinediaminemonohydrochloride,methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction