missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red, pure, high purity biological stain

Product Code. 10420891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420891 25 g Glass bottle 25g
10423265 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420891 Supplier Thermo Scientific Chemicals Supplier No. 229810250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-24-2
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.779
MDL Number MFCD00012651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Synonym Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride
PubChem CID 11105
ChEBI CHEBI:86370
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

Melting Point 290.0°C
Dye Content 50% min.
Beilstein 25, 401
Merck Index 15, 6573
Solubility Information Solubility in water: 50g/L. Other solubilities: 1.8% in abs. alcohol, 3.75% in cellosolve, 3.0% in ethylene glycol, practically insoluble in xylene
Infrared Spectrum Authentic
Formula Weight 288.77
Grade Pure
Packaging Glass bottle
Color Green
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Neutral red, High purity biological stain
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.