missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red, ACS

Product Code. 15464969
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15464969 50 g 50g
15474969 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464969 Supplier Thermo Scientific Chemicals Supplier No. J62643.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Nuclear counterstain useful in pH range 6.8 to 8.0 | CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol

Neutral Red is most commonly used for histological stain for lysosymes but has also been incorporated in investigation of viruses, as a pH indicator, determination of DNA with optical and electrochemical methods, evaluation of synthetic and naturally derived biomaterials. Neutral red can be electrochemically polymerized and used as a redox mediator. The compound once taken up by cells can be used to acertain cell viability with a excitation maximum at 470 nm and emission maximum at 580nm. It is used as a red nuclear counter stain in various cytochemical procedures. It is suitable for microscopy and microbiology and supravital staining of living blood cells. A vital dye used as an indicator and biological stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neutral Red is most commonly used for histological stain for lysosymes but has also been incorporated in investigation of viruses, as a pH indicator, determination of DNA with optical and electrochemical methods, evaluation of synthetic and naturally derived biomaterials. Neutral red can be electrochemically polymerized and used as a redox mediator. The compound once taken up by cells can be used to acertain cell viability with a excitation maximum at 470 nm and emission maximum at 580nm. It is used as a red nuclear counter stain in various cytochemical procedures. It is suitable for microscopy and microbiology and supravital staining of living blood cells. A vital dye used as an indicator and biological stain.

Solubility
Soluble in water (50 mg/ml at 25°C), abs. alcohol (1.8%), and cello solve (3.75%).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-24-2
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.779
MDL Number MFCD00012651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Synonym Basic Red 5; C.I. 50040
PubChem CID 11105
ChEBI CHEBI:86370
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

Melting Point 290°C (decomposition)
Beilstein 3918740
Merck Index 14,6488
Solubility Information Soluble in water (50mg/ml at 25°C),abs. alcohol (1.8%),and cello solve (3.75%).
Formula Weight 288.78
Grade ACS
Color Black to Green
Quantity 50 g
Physical Form Powder
Chemical Name or Material Neutral Red

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.