missing translation for 'onlineSavingsMsg'
Learn More

Neutral red, 60%, MP Biomedicals™

Product Code. 11473700 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473700 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473700 Supplier MP Biomedicals Supplier No. 0210243883
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 553-24-2
Assay Percent Range 0.6
Molecular Formula C15H17ClN4
Synonym Nuclear Fast Red, C.I. 50040
Color Green
Quantity 250 g
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Physical Form Powder
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
Molecular Weight (g/mol) 288.779
PubChem CID 11105
ChEBI CHEBI:86370
Formula Weight 288.8
Chemical Name or Material Neutral red
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.