missing translation for 'onlineSavingsMsg'
Learn More

Neutral Glass Specimen Tubes

Use these tubes for a variety of applications. Neutral Glass Specimen Tubes are ideal for samples which are susceptible to pH change.

Brand:  Samco T101/V1

Code : 02

Additional Details : Weight : 2.65000kg

 View more versions of this product

Product Code. 13130523

  • £606.00 / Pack of 882
In Stock
to see stock.
Add to basket

Description

Description

  • Tall specimen tubes
  • Neutral glass with screw neck
  • Good chemical resistance
  • Should not be taken from cold and placed directly into the hottest part of the flame without pre-heating
Specifications

Specifications

11 mm
1.75 mL
36 mm
Neutral Glass
882
882 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers