Learn More
Motic™ Neutral Density Filter, 45mm, BA210/BA400
Uniformly attenuate light intensity over a broad spectral range. Motic™ Neutral Density Filter, 45mm, BA210/BA400 reduces light equally with no resultant changes in color hue.
Brand:  Motic™ 1101001900361
Additional Details : Weight : 0.01000kg
Product Code. 15425202
  • £31.00 / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
  • For use with BA210/BA400 Microscopes.
  • Reduces or modifies the intensity of all wavelengths or colors of light equally, giving no changes in hue of color rendition.
Specifications
Specifications
Neutral Density
OD: 45mm
BA210/BA400 Microscopes