missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine™ SCF Mouse anti-Human, Clone: 1B3C4, Proteintech
Mouse Monoclonal Antibody
£122.57 - £303.85
Specifications
| Antigen | NeutraKine™ SCF |
|---|---|
| Clone | 1B3C4 |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
NeutraKine™ SCF Monoclonal antibody apecifically detetects NeutraKine™ SCF antigen in Human Samples. It is validated for Neutralization, ELISA.Specifications
| NeutraKine™ SCF | |
| Neutralization, ELISA | |
| Unconjugated | |
| Mouse | |
| Sterile PBS. | |
| KITLG | |
| IgG1 | |
| Protein G purification |
| 1B3C4 | |
| Monoclonal | |
| Liquid | |
| Human | |
| 4254 | |
| human Humankine SCF protein HZ-1024 | |
| Primary | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts