missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine™ IL-10 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£99.00 - £249.00
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | 1E4F5 |
| Applications | Immunohistochemistry, ELISA, Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233939
|
Proteintech
69018-1-IG-100UG |
100 μg |
£254.00 £249.00 / 100µg Save £5.00 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234890
|
Proteintech
69018-1-IG-25UG |
25 μg |
£101.00 £99.00 / 25µg Save £2.00 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeutraKine™ IL-10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™ IL-10. This antibody recognizes human antigen. The NeutraKine™ IL-10 antibody has been validated for the following applications: IHC, ELISA, Neutralization analysis.Specifications
| IL-10 | |
| Immunohistochemistry, ELISA, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| CSIF; IL-10; IL10A; MGC126450; MGC126451; TGIF | |
| IgG1 | |
| Protein G purification | |
| Neu Antibody |
| 1E4F5 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3586 | |
| RecombinantProtein | |
| Primary | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title