Learn More
NeutraKine FLT3 Ligand Mouse, anti-Human, Clone: 3E1A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69032-1-IG-25UG
Description
The NeutraKine FLT3 Ligand antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine FLT3 Ligand. This antibody recognizes human antigen. The NeutraKine FLT3 Ligand antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
| NeutraKine FLT3 Ligand | |
| 3E1A3 | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |
| ELISA, Neutralization | |
| FL, Flt3 ligand, Flt3L, FLT3LG, SL cytokine | |
| 25 μg | |
| 2323 | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.