Learn More
NeutraControl IL-12 p35 Mouse, anti-Human, Clone: 2C4G2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69529-1-IG-100UG
Description
The NeutraControl IL-12 p35 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraControl IL-12 p35. This antibody recognizes human antigen. The NeutraControl IL-12 p35 antibody has been validated for the following applications: Non-Neutralization, ELISA analysis.
Specifications
| NeutraControl IL-12 p35 | |
| 2C4G2 | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |
| ELISA | |
| CLMF, CLMF p35, IL 12 subunit p35, IL 12A, IL-12 p35, IL12A, Interleukin 12 subunit alpha, NFSK, NKSF1, P35 | |
| 100 μg | |
| 3592 | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.