missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neurotrypsin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557133
This item is not returnable.
View return policy
Description
Immunogen sequence: WVSVTDFGAP CLRWAEVPPF LERSPPASWA QLRGQRHNFC RSPDGAGRPW CFYGDARGKV DWGYCDCRHG SVRLRGGK Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.
Neurotrypsin is a central nervous system-expressed serine protease whose truncation or absence causes nonsyndromic mental retardation. It is most prominently expressed in structures that are involved in the processing and storage of learned behavior and memory, such as the cerebral cortex, the hippocampus, and amygdala. Evidence suggests that neurotrypsin has multiple functions, including axonal outgrowth, maintaining neuronal plasticity, and arranging the perineuronal environment, partly in coordination with other proteases including tissue plasminogen activator. At least two isoforms of neurotrypsin are known to exist.
Specifications
| Neurotrypsin | |
| Polyclonal | |
| Unconjugated | |
| PRSS12 | |
| Brain-specific serine protease 3; Bssp3; BSSP-3; leydin; Motopsin; MRT1; Neurotrypsin; Nt; peptidase, serine, 12 (neurotrypsin, motopsin); protease, serine 12; protease, serine 12 neurotrypsin (motopsin); protease, serine, 12 (neurotrypsin, motopsin); protease, serine, 12 neurotrypsin (motopsin); Prss12; Serine protease 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8492 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P56730 | |
| PRSS12 | |
| Recombinant protein corresponding to Human Neurotrypsin. Recombinant protein control fragment (Product #RP-102646). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction