missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurotrypsin Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76784MFV500
This item is not returnable.
View return policy
Description
Neurotrypsin Polyclonal antibody specifically detects Neurotrypsin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Neurotrypsin | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| brain-specific serine protease 3, BSSP3, BSSP-3, EC 3.4.21, leydin, MGC12722, Motopsin, MRT1EC 3.4.21.-, neurotrypsin, protease, serine, 12 (neurotrypsin, motopsin), Serine protease 12 | |
| Antibody was raised against a 23 amino acid synthetic peptide from near the amino terminus of human neurotrypsin. The immunogen is located within amino acids 110 - 160 of Neurotrypsin. Amino Acid Squence: VPPFLERSPPASWAQLRGQRHNF | |
| 0.1 mL | |
| Vision | |
| 8492 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction