missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurotrypsin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76784
This item is not returnable.
View return policy
Description
Neurotrypsin Polyclonal specifically detects Neurotrypsin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Neurotrypsin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| brain-specific serine protease 3, BSSP3, BSSP-3, EC 3.4.21, leydin, MGC12722, Motopsin, MRT1EC 3.4.21.-, neurotrypsin, protease, serine, 12 (neurotrypsin, motopsin), Serine protease 12 | |
| Rabbit | |
| 96 kDa | |
| 0.1 mg | |
| Vision | |
| 8492 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| CAA04816 | |
| PRSS12 | |
| Antibody was raised against a 23 amino acid synthetic peptide from near the amino terminus of human neurotrypsin. The immunogen is located within amino acids 110 - 160 of Neurotrypsin. Amino Acid Squence: VPPFLERSPPASWAQLRGQRHNF | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction