missing translation for 'onlineSavingsMsg'
Learn More

Neurotrimin/HNT Antibody (MM0485-5X25), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18424936 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424936 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424936 Supplier Novus Biologicals Supplier No. NBP211813JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neurotrimin/HNT Monoclonal antibody specifically detects Neurotrimin/HNT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurotrimin/HNT
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0485-5X25
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias hNT, IgLON family member 2, IGLON2HNTNT, MGC60329, neurotrimin, NTRI
Host Species Mouse
Immunogen Human recombinant Neurotrimin/HNT
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50863
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.