Learn More
Neuroplastin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA596882
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: QNEPRIVTSE EVIIRDSPVL PVTLQCNLTS SSHTLTYSYW TKNGVELSAT RKNASNMEYR INKPRAEDSG EYHCVYHFVS APKANATIEV KAAPDITGHK RSENKNEGQD ATMYCKSVGY PHPDWIWRKK ENGMPMDIVN TSGRFFIINK ENYTELNIVN LQITEDPGEY EC; Positive Samples: SKOV3, Jurkat; Cellular Location: Cell membrane, Single-pass type I membrane protein
This gene encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.Specifications
Neuroplastin | |
Polyclonal | |
Unconjugated | |
NPTN | |
NPTN | |
Recombit fusion protein containing a sequence corresponding to amino acids 29-200 of human NPTN. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
4.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97300, P97546, Q9Y639 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20320, 27020, 56064 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |