missing translation for 'onlineSavingsMsg'
Learn More

Neuroplastin 65 Antibody [DyLight 550], Novus Biologicals™

Product Code. 30735111 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735111 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735111 Supplier Novus Biologicals Supplier No. AF5360L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Neuroplastin 65 Polyclonal antibody specifically detects Neuroplastin 65 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroplastin 65
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias GP55;GP65;np55;np65;NPTN;SDFR1;SDR1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Neuroplastin 65, Gln29-His336, Accession # NP_036560
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27020
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.