missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropilin-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32241
This item is not returnable.
View return policy
Description
Neuropilin-2 Polyclonal specifically detects Neuropilin-2 in Human samples. It is validated for Western Blot.
Specifications
| Neuropilin-2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| MGC126574, neuropilin 2, neuropilin-2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17) | |
| Rabbit | |
| 105 kDa | |
| 0.1 mL | |
| Angiogenesis, Cancer, Hypoxia | |
| 8828 | |
| Human | |
| IgG |
| Western Blot | |
| 0.19mg/mL | |
| Western Blot 1:500-1:3000 | |
| O60462 | |
| NRP2 | |
| Recombinant protein encompassing a sequence within the center region of human NRP2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Predicted cross-reactivity based on sequence identity: Chicken (86%), Xenopus laevis (81%), and Xenopus tropicalis (82%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction