missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neuropilin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA129513
This item is not returnable.
View return policy
Description
This antibody recognizes a 130 kDa protein from mouse and rat brain tissue extract in western blot. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Neuropilin 1 is a 923 amino acid containing membrane bound coreceptor to a tyrosine kinase receptor belonging to the neuropilin family with two CUB domains, two F5/8 type C domains and a MAM domain. Neuropilin 1 forms a heterodimer with NRP2 (Neuropilin 2) and is known to bind to plexin (PLXNB1) to form semaphorin receptors. Expressed mostly in the nervous system this receptor is implicated in a wide range of functions including in the development of the cardiovascular system, angiogenesis, repulsive axon guidance, formation of certain neuronal circuits, cell survival, control of neuronal migration and organogenesis outside the nervous system. This semaphorin receptor functions as receptors for both the semaphorins and vascular endothelial growth factor and is also a regulator of VEGF-induced angiogenesis. Neuropilin 1 may be involved in aggressive advanced prostate carcinoma and in the progression of breast cancer.
Specifications
| Neuropilin 1 | |
| Polyclonal | |
| Unconjugated | |
| NRP1 | |
| A5 protein; BDCA4; C530029I03; CD304; Neuropilin; neuropilin 1; Neuropilin1; neuropilin-1; Neuropilin-1 precursor (A5 protein); NP1; NP-1; Npn1; NPN-1; NRP; NRP 1; Nrp1; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; vascular endothelial cell growth factor 165 receptor; VEGF165R | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18186, 246331, 8829 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| O14786, P97333, Q9QWJ9 | |
| NRP1 | |
| Within the range of amino acids 813-839 of rat Neuropilin 1. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction