missing translation for 'onlineSavingsMsg'
Learn More

Neuropilin-1 (CD304) Mouse anti-Human, Alexa Fluor 647, Clone: U21-1283, BD Pharmingen™

Mouse Monoclonal Antibody

£152.54 - £325.12

Specifications

Antigen Neuropilin-1 (CD304)
Clone U21-1283
Applications Flow Cytometry
Classification Monoclonal
Conjugate Alexa Fluor 647
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15884709
View Documents
BD Biosciences
566048
25 Tests
ONLINE EXCLUSIVE PRICE
£159.00
£152.54 / Each
Save £6.46
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15874709
View Documents
BD Biosciences
566047
100 Tests
ONLINE EXCLUSIVE PRICE
£339.00
£325.12 / Each
Save £13.88
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.

Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

Neuropilin-1 (CD304)
Flow Cytometry
Alexa Fluor 647
Mouse
Cell Biology
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Recombinant Human Neuropilin-1
Primary
Vol. Per test: 5μL
U21-1283
Monoclonal
Purified
RUO
Human
Neuropilin; NRP; NP1; Blood dendritic cell antigen 4; BDCA-4; VEGF165R
IgG2b κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.