missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurophysin II/Arg-vasopressin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93197-0.02ml
This item is not returnable.
View return policy
Description
Neurophysin II/Arg-vasopressin Polyclonal antibody specifically detects Neurophysin II/Arg-vasopressin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Neurophysin II/Arg-vasopressin | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| arginine vasopressin, ARVP, AVP-NPII, AVRP, neurohypophyseal, vasopressin-neurophysin 2-copeptin, VP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-164 of human AVP (NP_000481.2). CYFQNCPRGGKRAMSDLELRQCLPCGPGGKGRCFGPSICCADELGCFVGTAEALRCQEENYLPSPCQSGQKACGSGGRCAAFGVCCNDESCVTEPECREGFHRRARASDRSNATQLDGPAGALLLRLVQLAGAPEPFEPAQPDAY | |
| 0.02 mL | |
| Cell Biology, Lipid and Metabolism | |
| 551 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction