missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Neuropeptide Y (13-36), amide, human

Product Code. 16482919
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482919 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482919 Supplier MedChemExpress Supplier No. HYP1480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neuropeptide Y (13-36), amide, human is a selective neuropeptide Y2 receptor agonist.

Specifications

CAS 122341-40-6
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
For Use With (Application) Metabolism-protein/nucleotide metabolism
Molecular Weight (g/mol) 3000.4 g/mol
Product Type Peptides
Quantity 1 mg
Shipping Condition Room temperature
Synonym Neuropeptide Y (13-36), human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.