missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropeptide FF R1/NPFFR1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-168AF405
This item is not returnable.
View return policy
Description
Neuropeptide FF R1/NPFFR1 Polyclonal antibody specifically detects Neuropeptide FF R1/NPFFR1 in Human samples. It is validated for Western Blot
Specifications
| Neuropeptide FF R1/NPFFR1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 405 | |
| FLJ10751, G protein-coupled receptor 147, GPR147, neuropeptide FF 1, neuropeptide FF receptor 1, NPFF1G-protein coupled receptor 147, NPFF1R1, OT7T022, RFamide-related peptide receptor OT7T022 | |
| N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein. | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 64106 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction