missing translation for 'onlineSavingsMsg'
Learn More

Neuropathiazol, MedChemExpress

Product Code. 16460417
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460417 50 mg 50mg
16416767 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460417 Supplier Medchem Express Supplier No. HY10591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neuropathiazol, a neuronal differentiation inducer, selectively induces neuronal differentiation of multipotent hippocampal neural progenitor cells.

Neuropathiazol, a neuronal differentiation inducer, selectively induces neuronal differentiation of multipotent hippocampal neural progenitor cells.

Specifications

Chemical Name or Material Neuropathiazol
Physical Form Solid
CAS 880090-88-0
Color Brown
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H18N2O2S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 33.33 mg/mL (98.49 mM; Need ultrasonic)
SMILES CN(C1=CC=C(C(OCC)=O)C=C1)C2=CSC(C3=CC=CC=C3)=N2
Molecular Weight (g/mol) 338.42
Formula Weight 338.42
Percent Purity 98.27%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.