missing translation for 'onlineSavingsMsg'
Learn More

Neuronostatin Antibody (2956E) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30762563 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762563 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762563 Supplier Novus Biologicals Supplier No. FAB11554I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Neuronostatin Monoclonal antibody specifically detects Neuronostatin in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuronostatin
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2956E
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Neuronostatin-13, NST, preprosomatostatin, prepro-somatostatin
Host Species Rabbit
Immunogen Human Neuronostatin containing synthetic peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.