missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Neuronostatin-13 (human)

Product Code. 16422088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422088 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422088 Supplier MedChemExpress Supplier No. HYP1373
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Neuronostatin-13 human is a 13-amino acid peptide hormone encoded by the somatostatin gene and plays an important role in the regulation of hormonal and cardiac function.

Specifications

Purity 98.37%
CAS 1096485-24-3
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1415.68 g/mol
Product Type Peptides
Quantity 5 mg
Shipping Condition Room temperature
Solubility H2O : 25 mg/mL (17.66 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.