missing translation for 'onlineSavingsMsg'
Learn More

Neuronal Pentraxin 1 Antibody (OTI5G9), DyLight 594, Novus Biologicals™

Product Code. 18411503 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411503 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411503 Supplier Novus Biologicals Supplier No. NBP272965DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neuronal Pentraxin 1 Monoclonal antibody specifically detects Neuronal Pentraxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuronal Pentraxin 1
Applications Western Blot
Classification Monoclonal
Clone OTI5G9
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DKFZp686J2446, MGC105123, neuronal pentraxin INP1NP-I, neuronal pentraxin-1
Host Species Mouse
Immunogen Full length recombinant protein of human Neuronal Pentraxin 1 (NP_002513) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4884
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.