missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neuromedin U-8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101715
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NMU.
Neuromedin U (NmU) is a vertebrate peptide which stimulates uterine smooth muscle contraction and causes selective vasoconstriction. Like most other active peptides, it is proteolytically processed from a larger precursor protein. The mature peptides are 8 (NmU-8) to 25 (NmU-25) residues long and C-terminally amidated. The sequence of the C-terminal extremity of NmU is extremely well conserved across species.
Specifications
| Neuromedin U-8 | |
| Polyclonal | |
| Unconjugated | |
| NMU | |
| Neuromedin precursor-related peptide 33; Neuromedin precursor-related peptide 36; neuromedin U; neuromedin-U; Neuromedin-U-23; Neuromedin-U-25; Nmu; NmU 25; NmU-23; NmU-25; NURP33; NURP36; prepro-NMU | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10874 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P48645 | |
| NMU | |
| A synthesized peptide derived from human NMU(Accession P48645), corresponding to amino acid residues R143-I174. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction