missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurolysin Antibody (OTI1D6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72961PCP
This item is not returnable.
View return policy
Description
Neurolysin Monoclonal specifically detects Neurolysin in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| Neurolysin | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| NLN | |
| Full length recombinant protein of human Neurolysin (NP_065777) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI1D6 | |
| Flow Cytometry | |
| AGTBPFLJ23002, angiotensin binding protein, Angiotensin-binding protein, DKFZp564F123, EC 3.4.24, EC 3.4.24.16, KIAA1226EP24.16, MEP, mitochondrial, MOP, neurolysin (metallopeptidase M3 family) | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 57486.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction