missing translation for 'onlineSavingsMsg'
Learn More

Neurolysin Antibody (OTI1D6), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18407802 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407802 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407802 Supplier Novus Biologicals Supplier No. NBP272961MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neurolysin Monoclonal antibody specifically detects Neurolysin in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurolysin
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1D6
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias AGTBPFLJ23002, angiotensin binding protein, Angiotensin-binding protein, DKFZp564F123, EC 3.4.24, EC 3.4.24.16, KIAA1226EP24.16, MEP, mitochondrial, MOP, neurolysin (metallopeptidase M3 family)
Host Species Mouse
Immunogen Full length human recombinant protein of human NLN(NP_065777) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 57486
Target Species Human, Mouse, Rat, Primate, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.