missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroligin 4X/NLGN4X Antibody (S98-7), Novus Biologicals™
Mouse Monoclonal Antibody
£176.00 - £446.00
Specifications
| Antigen | Neuroligin 4X/NLGN4X |
|---|---|
| Clone | S98-7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664319
|
Novus Biologicals
NBP2-42198 |
0.1 mg |
£446.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648237
|
Novus Biologicals
NBP2-42198-0.025mg |
0.025 mg |
£176.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neuroligin 4X/NLGN4X Monoclonal antibody specifically detects Neuroligin 4X/NLGN4X in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Neuroligin 4X/NLGN4X | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Neuronal Cell Markers, Neuroscience | |
| PBS (pH 7.4), 50% Glycerol | |
| ASPGX2, HLNX, HNLX, KIAA1260AUTSX2, MGC22376, neuroligin 4, X-linked, NLGN, NLGN4neuroligin 4, X-linked | |
| Fusion protein amino acids 782-945 (intracellular C-terminus) of mouse Neuroligin 4. ∽45% identity with Neuroligin-1, -2 and -3. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S98-7 | |
| Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| B0F2B4 | |
| 57502 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title