missing translation for 'onlineSavingsMsg'
Learn More

Neuroligin 4X/NLGN4X Antibody [DyLight 350], Novus Biologicals™

Product Code. 30732772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732772 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732772 Supplier Novus Biologicals Supplier No. AF5158D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Neuroligin 4X/NLGN4X Polyclonal antibody specifically detects Neuroligin 4X/NLGN4X in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroligin 4X/NLGN4X
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ASPGX2, HLNX, HNLX, KIAA1260AUTSX2, MGC22376, neuroligin 4, X-linked, NLGN, NLGN4neuroligin 4, X-linked
Host Species Sheep
Immunogen E. coli-derived recombinant human Neuroligin 4/NLGN4, Met618-Thr673, Accession # Q8N0W4
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57502
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.