missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroligin 3/NLGN3 Antibody (242), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89824AF750
This item is not returnable.
View return policy
Description
Neuroligin 3/NLGN3 Monoclonal specifically detects Neuroligin 3/NLGN3 in Human samples. It is validated for Flow Cytometry.
Specifications
| Neuroligin 3/NLGN3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 242 | |
| Flow Cytometry | |
| ASPGX1, AUTSX1, Gliotactin homolog, KIAA1480HNL3, neuroligin 3, neuroligin-3, NL3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Neuroligin 3/NLGN3 (Q9NZ94-2; Met1-Ser689). | |
| 0.1 mL | |
| Neuroscience | |
| 54413 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction